Assay Details
Gene Symbol: Lrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008513.3 | NP_032539.2 | 4 - 5 | 1005 | 84 | |
GenBank mRNA | AF064984.1 | - | 4 - 5 | 995 | 84 | |
AF077847.1 | - | 4 - 5 | 883 | 84 | ||
AK147068.1 | - | 4 - 5 | 998 | 84 | ||
AK147231.1 | - | 5 - 6 | 1116 | 84 | ||
AK147330.1 | - | 5 - 6 | 1116 | 84 | ||
AK220186.1 | - | 4 - 5 | 958 | 84 | ||
BC011374.1 | - | 4 - 5 | 987 | 3154246 | 84 | |
BC094610.1 | - | 4 - 5 | 994 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
