Assay Details
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177629.1 | NP_001171100.1 | 3 - 4 | 367 | 94 | |
NM_010345.4 | NP_034475.2 | 3 - 4 | 527 | 94 | ||
GenBank mRNA | AF022072.1 | - | 3 - 4 | 501 | 94 | |
AK012646.1 | - | 3 - 4 | 507 | 94 | ||
AK030727.1 | - | 3 - 4 | 462 | 94 | ||
AK032927.1 | - | 3 - 4 | 508 | 94 | ||
AK163841.1 | - | 3 - 4 | 508 | 94 | ||
BC016111.1 | - | 3 - 4 | 367 | 4481345 | 94 | |
BC053842.1 | - | 3 - 4 | 494 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
