Assay Details
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177629.1 | NP_001171100.1 | 5 - 6 | 683 | 66 | |
NM_010345.4 | NP_034475.2 | 6 - 7 | 1008 | 66 | ||
GenBank mRNA | AF022072.1 | - | 6 - 7 | 982 | 66 | |
AK030727.1 | - | 6 - 7 | 943 | 66 | ||
AK163841.1 | - | 6 - 7 | 989 | 66 | ||
BC016111.1 | - | 5 - 6 | 683 | 4481345 | 66 | |
BC053842.1 | - | 6 - 7 | 975 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
