Assay Details
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177629.1 | NP_001171100.1 | 7 - 8 | 863 | 85 | |
NM_010345.4 | NP_034475.2 | 8 - 9 | 1188 | 85 | ||
GenBank mRNA | AF022072.1 | - | 8 - 9 | 1162 | 85 | |
AK030727.1 | - | 8 - 9 | 1123 | 85 | ||
AK032927.1 | - | 8 - 9 | 1169 | 85 | ||
AK163841.1 | - | 8 - 9 | 1169 | 85 | ||
AK169000.1 | - | 2 - 3 | 152 | 85 | ||
BC016111.1 | - | 7 - 8 | 863 | 4481345 | 85 | |
BC053842.1 | - | 8 - 9 | 1155 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
