Assay Details
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177629.1 | NP_001171100.1 | 9 - 10 | 1116 | 80 | |
NM_010345.4 | NP_034475.2 | 10 - 11 | 1441 | 80 | ||
GenBank mRNA | AF022072.1 | - | 10 - 11 | 1415 | 80 | |
AK030727.1 | - | 10 - 11 | 1376 | 80 | ||
AK032927.1 | - | 10 - 11 | 1422 | 80 | ||
AK163841.1 | - | 10 - 11 | 1422 | 80 | ||
AK169000.1 | - | 4 - 5 | 405 | 80 | ||
BC016111.1 | - | 9 - 10 | 1116 | 4481345 | 80 | |
BC053842.1 | - | 10 - 11 | 1408 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
