Assay Details
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011019.3 | NP_035149.2 | 9 - 10 | 1403 | 82 | |
GenBank mRNA | AB015978.1 | - | 9 - 10 | 2057 | 82 | |
AF058805.1 | - | 9 - 10 | 1373 | 82 | ||
AK157508.1 | - | 4 - 5 | 576 | 82 | ||
BC137703.1 | - | 8 - 9 | 1301 | 8860722 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
