Assay Details
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164071.1 | NP_001157543.1 | 3 - 4 | 464 | 84 | |
NM_011529.2 | NP_035659.1 | 2 - 3 | 255 | 84 | ||
GenBank mRNA | AK147727.1 | - | 3 - 4 | 410 | 84 | |
BC033282.1 | - | 2 - 3 | 191 | 84 | ||
U59864.1 | - | 2 - 3 | 268 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
