Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
TC0200000923.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
NM_001164071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
NM_001164071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
NM_001164072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
NM_001164072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
NM_011529.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007371.mm.1 |
NM_011529.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
TC0200000923.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
NM_001164071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
NM_001164071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
NM_001164072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
NM_001164072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
NM_011529.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007380.mm.1 |
NM_011529.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
TC0200000923.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
NM_001164071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
NM_001164071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
NM_001164072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
NM_001164072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
NM_011529.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007376.mm.1 |
NM_011529.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
TC0200000923.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
NM_001164071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
NM_001164071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
NM_001164072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
NM_001164072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
NM_011529.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007379.mm.1 |
NM_011529.2 |
Refseq |