Assay Details
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011678.2 | NP_035808.2 | 6 - 7 | 778 | 97 | |
GenBank mRNA | AK143582.1 | - | 6 - 7 | 713 | 97 | |
AK149964.1 | - | 6 - 7 | 736 | 97 | ||
AK169372.1 | - | 6 - 7 | 757 | 97 | ||
AK169933.1 | - | 6 - 7 | 748 | 97 | ||
AK171271.1 | - | 6 - 7 | 748 | 97 | ||
AK171633.1 | - | 6 - 7 | 731 | 97 | ||
AK220512.1 | - | 6 - 7 | 735 | 97 | ||
BC066180.1 | - | 6 - 7 | 688 | 97 | ||
BC066865.1 | - | 6 - 7 | 688 | 97 | ||
L00681.1 | - | 6 - 7 | 741 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
