Assay Details
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110791.1 | NP_001104261.1 | 11 - 12 | 1802 | 67 | |
NM_011750.2 | NP_035880.2 | 11 - 12 | 1802 | 67 | ||
NR_122118.1 | 10 - 11 | 1730 | 67 | |||
GenBank mRNA | AK133895.1 | - | 11 - 12 | 1593 | 67 | |
AK146151.1 | - | 11 - 12 | 1563 | 67 | ||
AK147081.1 | - | 11 - 12 | 1571 | 67 | ||
AK152914.1 | - | 10 - 11 | 1509 | 67 | ||
AK154705.1 | - | 11 - 12 | 1570 | 67 | ||
AK157845.1 | - | 11 - 12 | 1571 | 67 | ||
BC009091.1 | - | 11 - 12 | 1547 | 3157495 | 67 | |
BC055370.1 | - | 11 - 12 | 1561 | 30055109 | 67 | |
X80159.1 | - | 3 - 4 | 519 | 67 | ||
X85802.1 | - | 11 - 12 | 1559 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
