Assay Details
Gene Symbol: Tenm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011858.3 | NP_035988.2 | 11 - 12 | 2038 | 104 | |
GenBank mRNA | AB025413.1 | - | 9 - 10 | 1871 | 104 | |
AF059485.1 | - | 13 - 14 | 2476 | 104 | ||
AK147329.1 | - | 11 - 12 | 2039 | 104 | ||
AK147579.1 | - | 11 - 12 | 2338 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
