Assay Details
Gene Symbol: Ddb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015735.1 | NP_056550.1 | 16 - 17 | 2148 | 69 | |
GenBank mRNA | AB026432.1 | - | 16 - 17 | 2148 | 69 | |
AF159853.1 | - | 16 - 17 | 2108 | 69 | ||
AK145328.1 | - | 4 - 5 | 446 | 69 | ||
AK154303.1 | - | 16 - 17 | 2174 | 69 | ||
AK155020.1 | - | 16 - 17 | 2229 | 69 | ||
AK155920.1 | - | 16 - 17 | 2189 | 69 | ||
AK157491.1 | - | 16 - 17 | 2228 | 69 | ||
BC009661.1 | - | 16 - 17 | 2155 | 3487617 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
