Assay Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016692.3 | NP_057901.2 | 4 - 5 | 1204 | 94 | |
GenBank mRNA | AB100432.1 | - | 3 - 4 | 998 | 94 | |
AB100433.1 | - | 3 - 4 | 998 | 94 | ||
AF117610.1 | - | 4 - 5 | 1140 | 94 | ||
AK034764.1 | - | 4 - 5 | 1133 | 94 | ||
AK081841.1 | - | 4 - 5 | 1138 | 94 | ||
AK088627.1 | - | 4 - 5 | 1161 | 94 | ||
BC021761.2 | - | 4 - 5 | 1101 | 94 | ||
BC037011.1 | - | 4 - 5 | 1157 | 94 | ||
BC052414.1 | - | 4 - 5 | 1137 | 6401188 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
