Assay Details
Gene Symbol: Abcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018795.2 | NP_061265.2 | 27 - 28 | 3876 | 60 | |
GenBank mRNA | AB028737.1 | - | 27 - 28 | 3882 | 60 | |
BC040400.1 | - | 27 - 28 | 3884 | 60 | ||
BC049980.1 | - | 11 - 12 | 1819 | 60 | ||
EF109740.1 | - | 27 - 28 | 3876 | 60 | ||
EF109741.1 | - | 27 - 28 | 3876 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
