Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400010083.mm.1 |
TC0400001329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010083.mm.1 |
NM_001042411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010083.mm.1 |
NM_001042411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010083.mm.1 |
NM_001286148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010083.mm.1 |
NM_019783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010083.mm.1 |
NM_019783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
TC0400001329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
NM_001042411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
NM_001042411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
NM_001286148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
NM_019782.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
NM_019783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010080.mm.1 |
NM_019783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010085.mm.1 |
TC0400001329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010085.mm.1 |
NM_001042411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010085.mm.1 |
NM_001042411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010085.mm.1 |
NM_001286148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010085.mm.1 |
NM_019783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010085.mm.1 |
NM_019783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
TC0400001329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
NM_001042411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
NM_001042411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
NM_001286148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
NM_019782.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
NM_019783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010079.mm.1 |
NM_019783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010084.mm.1 |
TC0400001329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010084.mm.1 |
NM_001042411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010084.mm.1 |
NM_001042411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010084.mm.1 |
NM_001286148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010084.mm.1 |
NM_019783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010084.mm.1 |
NM_019783.2 |
Refseq |