Assay Details
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290509.1 | NP_001277438.1 | 1 - 2 | 260 | 75 | |
NM_020329.4 | NP_065062.1 | 1 - 2 | 260 | 75 | ||
NR_110963.1 | 1 - 2 | 260 | 75 | |||
GenBank mRNA | AB030189.1 | - | 1 - 2 | 100 | 75 | |
AK044141.1 | - | 1 - 2 | 99 | 75 | ||
AK053923.1 | - | 1 - 2 | 89 | 75 | ||
AK088065.1 | - | 1 - 2 | 104 | 75 | ||
BC026544.1 | - | 1 - 2 | 84 | 4975127 | 75 | |
BY133699.1 | - | 1 - 2 | 185 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102919 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102916 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102912 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102917 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102914 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102913 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102918 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990127 | AK088065.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990127 | BC026544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990127 | AK044141.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990127 | AK053923.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990127 | NM_020329.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682657 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820940 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113656 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150868 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990127 | AK088065.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990127 | BC026544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990127 | AK044141.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990127 | AK053923.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990127 | NM_020329.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246690 | AK088065.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246690 | BC026544.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246690 | AK010797.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246690 | AK044141.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246690 | AK053923.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102918 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102924 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102921 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102912 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102905 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00334163 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150868 | - | - |
Whole Mouse Genome G4122A | A_51_P246690 | AK088065.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P246690 | BC026544.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P246690 | AK010797.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P246690 | AK044141.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P246690 | AK053923.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118748 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138635 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150868 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02077602 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02077611 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02077614 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02077605 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102917 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102912 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102916 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102919 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102918 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102914 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02222282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01491405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01152357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01813687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01813689 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01591804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02077617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01813683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02077602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102970 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02068345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02077605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01565626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947589 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01565627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01813685 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01897092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01591805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21102969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02077611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02077614 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10471040 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471039 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471038 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471041 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471044 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471042 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471043 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471037 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428072_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10471036 | - | - |
Mouse Genome 430 2.0 Array | 1428072_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1428072_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57170_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57170_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200004303.mm.1 | TC0200000533.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200004303.mm.1 | NM_020329.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200004305.mm.1 | TC0200000533.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200004305.mm.1 | NM_020329.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200004300.mm.1 | TC0200000533.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200004300.mm.1 | NM_020329.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200004308.mm.1 | TC0200000533.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200004308.mm.1 | NM_020329.4 | Refseq |
Murine Genome U74v2 Set | 94373_PM_at | - | - |
Murine Genome U74v2 Set | 94373_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428072_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428072_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 877236 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6400053 | - | - |
MouseWG-6 v2 Expression BeadChip | 5080075 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6400053 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006400053 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718090 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723338 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2723338 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2718090 | - | - |
mouse-6 v1.1 (Illumina) | scl21089.8_496-S | - | - |
mouse-6 v1.1 (Illumina) | scl0003036.1_54-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5080075 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6400053 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl21089.8_496-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003036.1_54-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080075 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400053 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2966370 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2718090 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2723338 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2966370 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718090 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723338 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21089.8_496-S | - | - |
