Assay Details
Gene Symbol: Nelfcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020580.3 | NP_065605.2 | 7 - 8 | 889 | 70 | |
GenBank mRNA | AJ238741.1 | - | 7 - 8 | 823 | 70 | |
AJ238742.1 | - | 7 - 8 | 889 | 70 | ||
AJ238743.1 | - | 7 - 8 | 843 | 70 | ||
AK010360.1 | - | 7 - 8 | 836 | 70 | ||
AK049418.1 | - | 7 - 8 | 861 | 70 | ||
AK159866.1 | - | 7 - 8 | 890 | 70 | ||
AK187423.1 | - | 1 - 2 | 110 | 69 | ||
BC006959.1 | - | 7 - 8 | 850 | 3707929 | 70 | |
BC031747.1 | - | 7 - 8 | 847 | 3673038 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
