Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022321.4 | NP_071716.3 | 1 - 2 | 111 | 85 | |
GenBank mRNA | AF312712.1 | - | 1 - 2 | 69 | 85 | |
AK028329.1 | - | 1 - 2 | 112 | 85 | ||
AK037365.1 | - | 1 - 2 | 83 | 85 | ||
AK040914.1 | - | 1 - 2 | 42 | 85 | ||
AK079881.1 | - | 1 - 2 | 37 | 85 | ||
AK088419.1 | - | 1 - 2 | 61 | 85 | ||
BC011200.1 | - | 1 - 2 | 65 | 4190510 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF312712.1, AK028329.1, AK040914.1, AK079881.1, AK088419.1, BC011200.1, NM_022321.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
