Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162500.1 | NP_001155972.1 | 2 - 3 | 247 | 92 | |
NM_022321.4 | NP_071716.3 | 2 - 3 | 202 | 92 | ||
GenBank mRNA | AF312712.1 | - | 2 - 3 | 160 | 92 | |
AK037365.1 | - | 2 - 3 | 174 | 92 | ||
AK040914.1 | - | 2 - 3 | 133 | 92 | ||
AK079881.1 | - | 2 - 3 | 128 | 92 | ||
AK088419.1 | - | 2 - 3 | 152 | 92 | ||
BC011200.1 | - | 2 - 3 | 156 | 4190510 | 92 | |
BY200543.1 | - | 2 - 3 | 248 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
