Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162500.1 | NP_001155972.1 | 7 - 8 | 737 | 86 | |
NM_022321.4 | NP_071716.3 | 7 - 8 | 692 | 86 | ||
GenBank mRNA | AF312712.1 | - | 7 - 8 | 650 | 86 | |
AK028329.1 | - | 7 - 8 | 693 | 86 | ||
AK037365.1 | - | 7 - 8 | 665 | 86 | ||
AK040914.1 | - | 7 - 8 | 623 | 86 | ||
AK079881.1 | - | 7 - 8 | 618 | 86 | ||
AK088419.1 | - | 7 - 8 | 642 | 86 | ||
BC011200.1 | - | 7 - 8 | 646 | 4190510 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
