Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162500.1 | NP_001155972.1 | 9 - 10 | 813 | 69 | |
NM_022321.4 | NP_071716.3 | 9 - 10 | 768 | 69 | ||
GenBank mRNA | AF312712.1 | - | 9 - 10 | 726 | 69 | |
AK028329.1 | - | 9 - 10 | 769 | 69 | ||
AK037365.1 | - | 9 - 10 | 741 | 69 | ||
AK040914.1 | - | 9 - 10 | 699 | 69 | ||
AK079881.1 | - | 9 - 10 | 694 | 69 | ||
AK088419.1 | - | 9 - 10 | 718 | 69 | ||
BC011200.1 | - | 9 - 10 | 722 | 4190510 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
