Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025768.2 | NP_080044.2 | 2 - 3 | 289 | 85 | |
GenBank mRNA | AK007905.1 | - | 2 - 3 | 294 | 85 | |
AK017197.1 | - | 2 - 3 | 253 | 85 | ||
AK017251.1 | - | 2 - 3 | 200 | 85 | ||
AK044437.1 | - | 253 | 85 | |||
AK082863.1 | - | 2 - 3 | 283 | 85 | ||
AK149322.1 | - | 286 | 85 | |||
BC029096.1 | - | 2 - 3 | 295 | 3500563 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
