Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025768.2 | NP_080044.2 | 5 - 6 | 668 | 69 | |
GenBank mRNA | AF329833.1 | - | 3 - 4 | 456 | 69 | |
AK007905.1 | - | 5 - 6 | 673 | 69 | ||
AK017251.1 | - | 6 - 7 | 630 | 69 | ||
AK082863.1 | - | 5 - 6 | 662 | 69 | ||
AK149322.1 | - | 665 | 69 | |||
BC029096.1 | - | 5 - 6 | 674 | 3500563 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
