Assay Details
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025808.3 | NP_080084.2 | 9 - 10 | 1040 | 91 | |
GenBank mRNA | AK004561.2 | - | 9 - 10 | 1040 | 91 | |
AK005037.1 | - | 9 - 10 | 1039 | 91 | ||
AK083411.1 | - | 9 - 10 | 1044 | 91 | ||
AK132817.1 | - | 9 - 10 | 1039 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
