Assay Details
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025808.3 | NP_080084.2 | 17 - 18 | 2114 | 59 | |
GenBank mRNA | AK004561.2 | - | 17 - 18 | 2114 | 59 | |
AK005037.1 | - | 17 - 18 | 2113 | 59 | ||
AK083411.1 | - | 17 - 18 | 2118 | 59 | ||
AK088938.1 | - | 8 - 9 | 1046 | 59 | ||
AK132817.1 | - | 17 - 18 | 2113 | 59 | ||
AK208308.1 | - | 2 - 3 | 162 | 59 | ||
AK219434.1 | - | 2 - 3 | 163 | 59 | ||
BC034400.1 | - | 17 - 18 | 2108 | 2651456 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
