Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 2 - 3 | 207 | 90 | |
NM_026653.2 | NP_080929.1 | 2 - 3 | 207 | 90 | ||
GenBank mRNA | AK035084.1 | - | 2 - 3 | 135 | 90 | |
AK148035.1 | - | 2 - 3 | 148 | 90 | ||
AK150785.1 | - | 2 - 3 | 104 | 90 | ||
AK151590.1 | - | 2 - 3 | 160 | 90 | ||
AK152296.1 | - | 2 - 3 | 137 | 90 | ||
AK153107.1 | - | 2 - 3 | 137 | 90 | ||
AK153144.1 | - | 2 - 3 | 160 | 90 | ||
AK165316.1 | - | 2 - 3 | 120 | 90 | ||
AK165944.1 | - | 2 - 3 | 138 | 90 | ||
AK167598.1 | - | 2 - 3 | 99 | 90 | ||
AK168400.1 | - | 2 - 3 | 156 | 90 | ||
AK169599.1 | - | 2 - 3 | 137 | 90 | ||
AK197958.1 | - | 2 - 3 | 92 | 90 | ||
AK203654.1 | - | 2 - 3 | 92 | 90 | ||
BC019119.1 | - | 2 - 3 | 138 | 4487586 | 90 | |
CJ078143.1 | - | 2 - 3 | 207 | 90 | ||
CJ086677.1 | - | 2 - 3 | 207 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
