Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 14 - 15 | 1594 | 83 | |
NM_026653.2 | NP_080929.1 | 13 - 14 | 1531 | 83 | ||
GenBank mRNA | AK014298.1 | - | 2 - 3 | 234 | 83 | |
AK019864.1 | - | 7 - 8 | 932 | 83 | ||
AK035084.1 | - | 13 - 14 | 1459 | 83 | ||
AK148035.1 | - | 13 - 14 | 1472 | 83 | ||
AK150785.1 | - | 13 - 14 | 1428 | 83 | ||
AK151590.1 | - | 13 - 14 | 1484 | 83 | ||
AK152296.1 | - | 14 - 15 | 1524 | 83 | ||
AK153107.1 | - | 14 - 15 | 1524 | 83 | ||
AK153144.1 | - | 13 - 14 | 1484 | 83 | ||
AK165316.1 | - | 13 - 14 | 1444 | 83 | ||
AK165944.1 | - | 13 - 14 | 1462 | 83 | ||
AK167598.1 | - | 13 - 14 | 1417 | 83 | ||
AK168400.1 | - | 13 - 14 | 1484 | 83 | ||
AK169599.1 | - | 13 - 14 | 1455 | 83 | ||
AK207399.1 | - | 2 - 3 | 270 | 83 | ||
BC019119.1 | - | 13 - 14 | 1462 | 4487586 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
