Assay Details
Gene Symbol: Pask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080850.2 | NP_543126.2 | 16 - 17 | 4155 | 75 | |
GenBank mRNA | AK028435.1 | - | 16 - 17 | 4157 | 75 | |
AK081074.1 | - | 858 | 75 | |||
AK083528.1 | - | 16 - 17 | 4127 | 75 | ||
AK083590.1 | - | 16 - 17 | 4121 | 75 | ||
AK129063.1 | - | 16 - 17 | 4045 | 75 | ||
AK141220.1 | - | 792 | 75 | |||
BC029725.1 | - | 1 - 2 | 350 | 75 | ||
BC150821.1 | - | 15 - 16 | 3867 | 9087732 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
