Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080853.3 | NP_543129.3 | 9 - 10 | 1979 | 78 | |
GenBank mRNA | AF324864.1 | - | 9 - 10 | 1812 | 78 | |
AK043753.1 | - | 8 - 9 | 1432 | 78 | ||
AK045409.1 | - | 9 - 10 | 1742 | 78 | ||
BC038375.1 | - | 9 - 10 | 1358 | 5357143 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
