Assay Details
Gene Symbol: Recql5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130454.2 | NP_569721.1 | 10 - 11 | 2162 | 72 | |
GenBank mRNA | AB059999.1 | - | 10 - 11 | 1676 | 72 | |
AK049269.1 | - | 5 - 6 | 562 | 72 | ||
AK050965.1 | - | 8 - 9 | 891 | 72 | ||
BC023769.1 | - | 9 - 10 | 1454 | 72 | ||
BC053086.1 | - | 10 - 11 | 2415 | 72 | ||
BC059000.1 | - | 10 - 11 | 2135 | 72 | ||
BC060070.1 | - | 10 - 11 | 2415 | 72 | ||
BC066176.1 | - | 10 - 11 | 1885 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
