Assay Details
Gene Symbol: Cdhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130878.2 | NP_570948.1 | 7 - 8 | 813 | 65 | |
GenBank mRNA | AB093300.1 | - | 7 - 8 | 1229 | 65 | |
AF426393.1 | - | 7 - 8 | 700 | 65 | ||
AK143151.1 | - | 7 - 8 | 753 | 66 | ||
BC042454.1 | - | 7 - 8 | 723 | 5369576 | 65 | |
BC058270.1 | - | 7 - 8 | 813 | 6490597 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
