Assay Details
Gene Symbol: Arf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007480.1 | NP_031506.1 | 5 - 6 | 505 | 97 | |
GenBank mRNA | BC132452.1 | - | 6 - 7 | 493 | 40130729 | 97 |
BC138756.1 | - | 6 - 7 | 493 | 8861778 | 97 | |
D87902.1 | - | 5 - 6 | 505 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067445 | NM_007480.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067445 | D87902.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976207 | NM_007480.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976207 | BC132452.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976207 | BC138756.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976207 | D87902.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204093 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067445 | NM_007480.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067445 | D87902.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976207 | NM_007480.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976207 | BC132452.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976207 | BC138756.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976207 | D87902.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381801 | NM_007480.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381801 | BC132452.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381801 | BC138756.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381801 | D87902.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320128 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320115 | - | - |
Whole Mouse Genome G4122A | A_51_P381801 | NM_007480.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P381801 | BC132452.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P381801 | BC138756.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P381801 | D87902.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05694145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05058249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05694148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05694157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24320103 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10536748 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10536749 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10536751 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10536750 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10536747 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10536746 | - | - |
Mouse Genome 430 2.0 Array | 1422807_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422807_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11844_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11844_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600001353.mm.1 | TC0600000213.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600001353.mm.1 | NM_007480.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600001353.mm.1 | NM_007480.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600001352.mm.1 | TC0600000213.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600001352.mm.1 | NM_007480.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600001352.mm.1 | NM_007480.1 | Refseq |
Mouse Expression Set 430 | 1422807_at | - | - |
Mouse Expression Set 430 | 1422807_PM_at | - | - |
Murine Genome U74v2 Set | 92968_at | - | - |
Murine Genome U74v2 Set | 92968_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422807_at | - | - |
Mouse Genome 430A 2.0 Array | 1422807_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 566025 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4730369 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4730369 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004730369 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2677712 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2677712 | - | - |
mouse-6 v1.1 (Illumina) | scl011844.1_12-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4730369 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl011844.1_12-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730369 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2677712 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2677712 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl011844.1_12-S | - | - |
