Assay Details
Gene Symbol: Fech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007998.6 | NP_032024.2 | 7 - 8 | 899 | 91 | |
GenBank mRNA | AK004718.2 | - | 7 - 8 | 900 | 91 | |
AK018738.2 | - | 7 - 8 | 902 | 91 | ||
AK084843.1 | - | 7 - 8 | 797 | 91 | ||
AK150685.1 | - | 7 - 8 | 901 | 91 | ||
AK159652.1 | - | 7 - 8 | 932 | 91 | ||
AK189170.1 | - | 3 - 4 | 309 | 91 | ||
AK199138.1 | - | 3 - 4 | 309 | 91 | ||
BC006746.1 | - | 7 - 8 | 906 | 3256604 | 91 | |
BC094548.1 | - | 7 - 8 | 872 | 91 | ||
BC096770.1 | - | 6 - 7 | 733 | 91 | ||
M59288.1 | - | 6 - 7 | 736 | 91 | ||
M61215.1 | - | 7 - 8 | 886 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
