Assay Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 11 - 12 | 1794 | 72 | |
GenBank mRNA | AK157842.1 | - | 11 - 12 | 1807 | 72 | |
AK166179.1 | - | 11 - 12 | 1782 | 72 | ||
BC047117.1 | - | 11 - 12 | 1802 | 4951173 | 72 | |
BC050897.1 | - | 11 - 12 | 1777 | 6401835 | 72 | |
U21011.1 | - | 11 - 12 | 1811 | 72 | ||
X81143.1 | - | 11 - 12 | 1794 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
