Assay Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 14 - 15 | 2499 | 70 | |
GenBank mRNA | AK157842.1 | - | 14 - 15 | 2512 | 70 | |
AK166179.1 | - | 14 - 15 | 2488 | 70 | ||
BC047117.1 | - | 14 - 15 | 2507 | 4951173 | 70 | |
BC050897.1 | - | 14 - 15 | 2482 | 6401835 | 70 | |
U21011.1 | - | 14 - 15 | 2516 | 70 | ||
X81143.1 | - | 14 - 15 | 2499 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
