Assay Details
Gene Symbol: Ncoa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008679.3 | NP_032705.2 | 3 - 4 | 311 | 64 | |
GenBank mRNA | AF000581.1 | - | 2 - 3 | 200 | 64 | |
AK137507.1 | - | 3 - 4 | 297 | 64 | ||
AK142306.1 | - | 3 - 4 | 216 | 64 | ||
AK156345.1 | - | 3 - 4 | 289 | 64 | ||
BC086482.1 | - | 3 - 4 | 281 | 30104508 | 64 | |
BC141177.1 | - | 2 - 3 | 142 | 9055789 | 64 | |
CJ101424.1 | - | 3 - 4 | 312 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
