Assay Details
Gene Symbol: Ncoa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008679.3 | NP_032705.2 | 11 - 12 | 1703 | 67 | |
GenBank mRNA | AF000581.1 | - | 10 - 11 | 1595 | 67 | |
AK137507.1 | - | 11 - 12 | 1689 | 67 | ||
AK142306.1 | - | 11 - 12 | 1608 | 67 | ||
AK156345.1 | - | 11 - 12 | 1681 | 67 | ||
BC086482.1 | - | 11 - 12 | 1673 | 30104508 | 67 | |
BC093478.1 | - | 8 - 9 | 1334 | 67 | ||
BC141177.1 | - | 10 - 11 | 1534 | 9055789 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
