Assay Details
Gene Symbol: Npas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008719.2 | NP_032745.2 | 15 - 16 | 1986 | 69 | |
GenBank mRNA | BC109166.2 | - | 15 - 16 | 1986 | 40048145 | 69 |
U77969.1 | - | 15 - 16 | 2060 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
