Assay Details
Gene Symbol: Ptpn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008979.2 | NP_033005.1 | 4 - 5 | 494 | 128 | |
NR_104070.1 | 6 - 7 | 519 | 128 | |||
GenBank mRNA | AK153812.1 | - | 6 - 7 | 520 | 128 | |
M90388.1 | - | 4 - 5 | 479 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
