Assay Details
Gene Symbol: Ptpn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008979.2 | NP_033005.1 | 16 - 17 | 2171 | 65 | |
NR_104070.1 | 18 - 19 | 2196 | 65 | |||
GenBank mRNA | AK153812.1 | - | 18 - 19 | 2197 | 65 | |
BC055377.1 | - | 16 - 17 | 2093 | 30077916 | 65 | |
M90388.1 | - | 16 - 17 | 2156 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
