Assay Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286260.1 | NP_001273189.1 | 5 - 6 | 1503 | 111 | |
NM_018823.2 | NP_061293.2 | 6 - 7 | 1557 | 111 | ||
NM_133957.3 | NP_598718.2 | 7 - 8 | 1619 | 111 | ||
GenBank mRNA | AF162853.1 | - | 5 - 6 | 1414 | 111 | |
AF200687.1 | - | 6 - 7 | 1192 | 111 | ||
AF228705.1 | - | 3 - 4 | 1257 | 111 | ||
AF228706.1 | - | 3 - 4 | 1257 | 111 | ||
AF369980.1 | - | 7 - 8 | 1258 | 111 | ||
AK134022.1 | - | 7 - 8 | 1623 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
