Assay Details
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB028920.1 | - | 11 - 12 | 1430 | 76 | |
AK004801.1 | - | 11 - 12 | 1436 | 76 | ||
AK089092.1 | - | 11 - 12 | 1443 | 76 | ||
BC042602.1 | - | 11 - 12 | 2313 | 76 | ||
BC054433.1 | - | 9 - 10 | 2010 | 76 | ||
BC060170.1 | - | 11 - 12 | 2310 | 76 | ||
BC060209.1 | - | 10 - 11 | 1926 | 76 | ||
BC078448.1 | - | 10 - 11 | 1816 | 76 | ||
BC125521.1 | - | 11 - 12 | 1426 | 40130036 | 76 | |
BC137746.1 | - | 11 - 12 | 1426 | 8860765 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
