Assay Details
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277292.1 | NP_001264221.1 | 18 - 19 | 2646 | 96 | |
NM_001277293.1 | NP_001264222.1 | 19 - 20 | 2802 | 96 | ||
NM_001277294.1 | NP_001264223.1 | 18 - 19 | 2503 | 96 | ||
NM_028736.2 | NP_083012.1 | 19 - 20 | 2659 | 96 | ||
NM_130891.2 | NP_570961.1 | 17 - 18 | 2422 | 96 | ||
NM_133442.2 | NP_597699.1 | 17 - 18 | 2565 | 96 | ||
GenBank mRNA | AB051560.1 | - | 17 - 18 | 2299 | 96 | |
AB051561.1 | - | 19 - 20 | 2536 | 96 | ||
AB051562.1 | - | 17 - 18 | 2285 | 96 | ||
AK220553.1 | - | 18 - 19 | 2360 | 96 | ||
BC067398.1 | - | 18 - 19 | 2503 | 30096759 | 96 | |
BC072632.1 | - | 19 - 20 | 2802 | 6843802 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
