Assay Details
Gene Symbol: Nt5c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164363.1 | NP_001157835.1 | 10 - 11 | 1275 | 91 | |
NM_001164365.1 | NP_001157837.1 | 10 - 11 | 932 | 91 | ||
NM_029810.4 | NP_084086.3 | 10 - 11 | 863 | 91 | ||
NR_028353.1 | 11 - 12 | 922 | 91 | |||
GenBank mRNA | AK008045.1 | - | 10 - 11 | 810 | 91 | |
AK030786.1 | - | 11 - 12 | 881 | 91 | ||
AK036730.1 | - | 10 - 11 | 821 | 91 | ||
AK038458.1 | - | 7 - 8 | 569 | 91 | ||
AK132384.1 | - | 10 - 11 | 821 | 91 | ||
AK147901.1 | - | 10 - 11 | 821 | 91 | ||
AK162385.1 | - | 10 - 11 | 1240 | 91 | ||
AK169616.1 | - | 10 - 11 | 821 | 91 | ||
BC064760.1 | - | 9 - 10 | 797 | 30436992 | 91 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BC006028.1 | - | Chr. - - [-] on Build | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
