Assay Details
Gene Symbol: Dcaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165256.1 | NP_001158728.1 | 1 - 2 | 192 | 140 | |
NM_030246.2 | NP_084522.1 | 2 - 3 | 249 | 140 | ||
GenBank mRNA | BB852314.1 | - | 2 - 3 | 170 | 140 | |
BC003284.1 | - | 1 - 2 | 158 | 3582095 | 140 | |
CN722277.1 | - | 1 - 2 | 167 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC003284.1, NM_030246.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
