Assay Details
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080555.2 | NP_542122.1 | 3 - 4 | 1057 | 58 | |
GenBank mRNA | AK011276.1 | - | 3 - 4 | 1056 | 58 | |
AK076183.1 | - | 3 - 4 | 753 | 58 | ||
AK160056.1 | - | 3 - 4 | 957 | 58 | ||
BC005558.1 | - | 3 - 4 | 1044 | 3964817 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
