Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133681.4 | NP_598442.1 | 3 - 4 | 400 | 73 | |
GenBank mRNA | AK078869.1 | - | 3 - 4 | 401 | 73 | |
AK162521.1 | - | 3 - 4 | 400 | 73 | ||
BC003448.1 | - | 3 - 4 | 340 | 2654254 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
