Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133681.4 | NP_598442.1 | 4 - 5 | 482 | 61 | |
GenBank mRNA | AK078869.1 | - | 4 - 5 | 483 | 61 | |
AK162521.1 | - | 4 - 5 | 482 | 61 | ||
BC003448.1 | - | 4 - 5 | 422 | 2654254 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
