Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300020626.mm.1 |
TC0300002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020626.mm.1 |
NM_001159354.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020626.mm.1 |
NM_001159354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020626.mm.1 |
NM_133853.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020626.mm.1 |
NM_133853.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020623.mm.1 |
TC0300002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020623.mm.1 |
NM_001159354.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020623.mm.1 |
NM_001159354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020623.mm.1 |
NM_133853.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020623.mm.1 |
NM_133853.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020624.mm.1 |
TC0300002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020624.mm.1 |
NM_001159354.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020624.mm.1 |
NM_001159354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020624.mm.1 |
NM_133853.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020624.mm.1 |
NM_133853.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020622.mm.1 |
TC0300002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020622.mm.1 |
NM_001159354.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020622.mm.1 |
NM_001159354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020622.mm.1 |
NM_133853.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020622.mm.1 |
NM_133853.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020625.mm.1 |
TC0300002636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020625.mm.1 |
NM_001159354.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020625.mm.1 |
NM_001159354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020625.mm.1 |
NM_133853.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020625.mm.1 |
NM_133853.3 |
Refseq |