Assay Details
Gene Symbol: Magi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159354.1 | NP_001152826.1 | 4 - 5 | 1214 | 101 | |
NM_133853.3 | NP_598614.1 | 4 - 5 | 1214 | 101 | ||
GenBank mRNA | AF213258.1 | - | 4 - 5 | 835 | 101 | |
AK030083.1 | - | 4 - 5 | 618 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
